2-Methyl-1-(2-pyridyl)-1-propylamine 98%

Supplier: Ambeed
Warning

A638843-10G A638843-1G A638843-5G A638843-250MG
77018-268EA 299 USD
77018-268 77018-270 77018-272 77891-271
2-Methyl-1-(2-pyridyl)-1-propylamine 98%
2-Methyl-1-(2-pyridyl)-1-propylamine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MDL Number: MFCD09802325
CAS Number: 58088-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR