2-Bromobenzo[d]thiazol-6-amine 95%
Supplier: Ambeed
|
Warning
|
A637883-100MG
A637883-1G
A637883-250MG
77035-376EA
32.89
USD
77035-376
77035-378
77035-380
2-Bromobenzo[d]thiazol-6-amine 95%
2-Bromobenzo[d]thiazol-6-amine
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂S MW: 229.10 g/mol |
MDL Number:
MFCD11977496 CAS Number: 945400-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...