N-(3-Chloro-4-fluorophenyl)-7-(3-morpholinopropoxy)-6-nitroquinazolin-4-amine 95%

Supplier: Ambeed
Warning

A637607-100MG A637607-1G A637607-250MG A637607-50MG
76992-284EA 159.14 USD
76992-284 76992-286 76992-288 76992-290
N-(3-Chloro-4-fluorophenyl)-7-(3-morpholinopropoxy)-6-nitroquinazolin-4-amine 95%
N-(3-Chloro-4-fluorophenyl)-7-(3-morpholinopropoxy)-6-nitroquinazolin-4-amine

Precaución: for Research Use Only

Formula: C₂₁H₂₁ClFN₅O₄
MW: 461.87 g/mol
MDL Number: MFCD12032146
CAS Number: 267243-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR