3-Bromo-6-nitroimidazo[1,2-a]pyridine 98%

Supplier: Ambeed
Warning

A636028-100MG A636028-1G A636028-250MG A636028-5G
77005-636EA 14.86 USD
77005-636 77005-638 77005-640 77569-218
3-Bromo-6-nitroimidazo[1,2-a]pyridine 98%
3-Bromo-6-nitroimidazo[1,2-a]pyridine

Precaución: For research use only.

Formula: C₇H₄BrN₃O₂
MW: 242.0296 g/mol
MDL Number: MFCD13193088
CAS Number: 52310-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR