Methyl 3-(benzyloxy)cyclobutanecarboxylate 95%

Supplier: Ambeed
Warning

A635518-1G A635518-250MG A635518-5G
77035-354EA 103.97 USD
77035-354 77035-356 77035-358
Methyl 3-(benzyloxy)cyclobutanecarboxylate 95%
Methyl 3-(benzyloxy)cyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₃
MW: 220.26 g/mol
MDL Number: MFCD00513559
CAS Number: 4934-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR