(1R,2S)-2-Phenylcyclopropanamine 95%

Supplier: Ambeed
Warning

A634641-100MG A634641-250MG A634641-1G
76992-270EA 76.39 USD
76992-270 76992-272 77613-958
(1R,2S)-2-Phenylcyclopropanamine 95%
(1R,2S)-2-Phenylcyclopropanamine

Precaución: for Research Use Only

Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD00865363
CAS Number: 3721-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR