1-Ethyl-1H-indole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A634548-1G A634548-250MG A634548-5G
77018-242EA 173 USD
77018-242 77018-244 77018-246
1-Ethyl-1H-indole-2-carbaldehyde 95%
1-Ethyl-1H-indole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO
MDL Number: MFCD11877851
CAS Number: 40913-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR