(R)-1-(2-Fluoro-4-methoxyphenyl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A633415-250MG A633415-100MG A633415-1G
76992-268EA 62 USD
76992-268 77891-265 77891-266
(R)-1-(2-Fluoro-4-methoxyphenyl)ethanamine hydrochloride 95%
(R)-1-(2-Fluoro-4-methoxyphenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClFNO
MDL Number: MFCD16293363
CAS Number: 1309598-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR