Ethyl 5-amino-2-fluorobenzoate 98%

Supplier: Ambeed
Warning

A632805-1G A632805-5G
77018-224EA 6 USD
77018-224 77018-226
Ethyl 5-amino-2-fluorobenzoate 98%
Ethyl 5-amino-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MDL Number: MFCD09037896
CAS Number: 123207-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR