N-tert-Butyl 4-boronobenzenesulfonamide pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide

A632747-1G A632747-250MG A632747-5G
77018-218EA 96.55 USD
77018-218 77018-220 77018-222
N-tert-Butyl 4-boronobenzenesulfonamide pinacol ester 98%
N-tert-Butyl 4-boronobenzenesulphonamide pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₆BNO₄S
MW: 339.2579 g/mol
MDL Number: MFCD06657804
CAS Number: 648905-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR