2-Amino-3-chloro-5-nitropyridine 98%

Supplier: Ambeed
Warning

A632657-10G A632657-1G A632657-250MG A632657-25G
77005-618EA 43 USD
77005-618 77005-620 77005-622 77005-624
2-Amino-3-chloro-5-nitropyridine 98%
2-Amino-3-chloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄ClN₃O₂
MW: 173.5600 g/mol
MDL Number: MFCD00160299
CAS Number: 22353-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR