5-Ethyl-2,4-dihydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A632623-100MG A632623-1G A632623-250MG
77018-212EA 96.55 USD
77018-212 77018-214 77018-216
5-Ethyl-2,4-dihydroxybenzaldehyde 95%
5-Ethyl-2,4-dihydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.17 g/mol
MDL Number: MFCD21606577
CAS Number: 37470-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR