2-Bromo-4-methylbenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-methyl-1-(trifluoromethyl)benzene

A632160-1G A632160-250MG
77005-614EA 56.23 USD
77005-614 77005-616
2-Bromo-4-methylbenzotrifluoride 98%
2-Bromo-4-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD11226554
CAS Number: 121793-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR