meso-1,2-Bis(4-chlorophenyl)ethylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: (1R,2S)-rel-1,2-Bis(4-chlorophenyl)ethane-1,2-diamine

A631851-100MG A631851-1G A631851-250MG A631851-5G
77018-204EA 23.34 USD
77018-204 77018-206 77018-208 77018-210
meso-1,2-Bis(4-chlorophenyl)ethylenediamine 98%
meso-1,2-Bis(4-chlorophenyl)ethylenediamine

Precaución: for Research Use Only

Formula: C₁₄H₁₄Cl₂N₂
MW: 281.1804 g/mol
MDL Number: MFCD05150371
CAS Number: 74641-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR