(R)-Methyl 2-amino-3-(1H-indol-3-yl)propanoate 97%

Supplier: Ambeed
Warning

A631025-5G A631025-25G
77018-202EA 14.86 USD
77018-202 77684-516
(R)-Methyl 2-amino-3-(1H-indol-3-yl)propanoate 97%
(R)-Methyl 2-amino-3-(1H-indol-3-yl)propanoate

Precaución: For research use only.

Formula: C₁₂H₁₄N₂O₂
MW: 218.25 g/mol
MDL Number: MFCD08669786
CAS Number: 22032-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR