(3-Bromopyridin-4-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A630670-100MG A630670-1G
77018-198EA 79 USD
77018-198 77018-200
(3-Bromopyridin-4-yl)methanamine hydrochloride 95%
(3-Bromopyridin-4-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₈BrClN₂
MDL Number: MFCD27923663
CAS Number: 1956307-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR