2-(3-Aminopropyl)isoindoline-1,3-dione hydrochloride 97%

Supplier: Ambeed
Warning

A630413-250MG A630413-1G A630413-5G
77018-196EA 20 USD
77018-196 77891-248 77891-249
2-(3-Aminopropyl)isoindoline-1,3-dione hydrochloride 97%
2-(3-Aminopropyl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂O₂
MW: 240.68 g/mol
MDL Number: MFCD00035196
CAS Number: 121821-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR