Methyl 2-chloro-3-hydroxybenzoate 95%

Supplier: Ambeed
Warning

A629444-100MG A629444-250MG A629444-5G A629444-1G
76992-248EA 39.26 USD
76992-248 76992-250 76992-252 77684-514
Methyl 2-chloro-3-hydroxybenzoate 95%
Methyl 2-chloro-3-hydroxybenzoate

Precaución: For research use only.

Formula: C₈H₇ClO₃
MW: 186.592 g/mol
MDL Number: MFCD13189670
CAS Number: 1125632-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR