Ethyl-2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-6-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-6-carboxylate

A629431-1G A629431-250MG
77005-582EA 83.82 USD
77005-582 77005-584
Ethyl-2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-6-carboxylate 98%
Ethyl-2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂S
MW: 226.3 g/mol
MDL Number: MFCD13195375
CAS Number: 134136-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR