Dibenzo[b,d]furan-4-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Dibenzofuran-4-carboxaldehyde

A629428-1G A629428-25G A629428-5G
77018-190EA 26 USD
77018-190 77018-192 77018-194
Dibenzo[b,d]furan-4-carbaldehyde 98%
Dibenzo[b,d]furan-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈O₂
MDL Number: MFCD03306027
CAS Number: 96706-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR