(3-Chloro-6-methoxypyridin-2-yl)methanamine 95%

Supplier: Ambeed
Warning

A629150-1G A629150-250MG
77018-180EA 704.44 USD
77018-180 77018-182
(3-Chloro-6-methoxypyridin-2-yl)methanamine 95%
(3-Chloro-6-methoxypyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
MW: 172.61 g/mol
MDL Number: MFCD13189329
CAS Number: 1060810-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR