tert-Butyl ((2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl)carbamate succinate(2:1) 98+%

Supplier: Ambeed
Warning

A628996-25G A628996-5G
77005-578EA 38 USD
77005-578 77005-580
tert-Butyl ((2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl)carbamate succinate(2:1) 98+%
tert-Butyl ((2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl)carbamate succinate(2:1)

Precaución: for Research Use Only

Formula: C₅₀H₇₀N₄O₁₀
MDL Number: MFCD11041076
CAS Number: 183388-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR