(S)-(-)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(-)-2-(Diphenylphosphino)-2'-methoxy-1,1'-binaphthyl, (S)-(2'-Methoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine, (S)-(-)-MOP, (S)-MOP
A628791-1G
A628791-250MG
77005-574EA
544.25
USD
77005-574
77005-576
(S)-(-)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl 98%
(S)-(-)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl
Precaución: for Research Use Only
Formula:
C₃₃H₂₅OP MW: 468.536 g/mol |
MDL Number:
MFCD00269686 CAS Number: 134484-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...