5-Chlorobenzo[c]isoxazole 95%

Supplier: Ambeed
Warning

A628631-1G A628631-250MG
76992-244EA 243 USD
76992-244 77891-244
5-Chlorobenzo[c]isoxazole 95%
5-Chlorobenzo[c]isoxazole

Precaución: for Research Use Only

Formula: C₇H₄ClNO
MDL Number: MFCD04004099
CAS Number: 4596-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR