Sodium pyridine-3-sulfinate 97%

Supplier: Ambeed
Warning

A627805-1G A627805-250MG
77018-166EA 175 USD
77018-166 77018-168
Sodium pyridine-3-sulfinate 97%
Sodium pyridine-3-sulfinate

Precaución: for Research Use Only

Formula: C₅H₄NNaO₂S
MDL Number: MFCD24113335
CAS Number: 123151-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR