2-Methoxy-5-(trifluoromethyl)phenol 98%

Supplier: Ambeed
Danger

A627790-1G A627790-250MG
77018-162EA 79 USD
77018-162 77018-164
2-Methoxy-5-(trifluoromethyl)phenol 98%
2-Methoxy-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₈H₇F₃O₂
MDL Number: MFCD09832299
CAS Number: 349-67-7
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR