tert-Butyl (2-amino-2-iminoethyl)carbamate hydrochloride 95%

Supplier: Ambeed
Warning

A627256-1G A627256-250MG A627256-5G
76992-238EA 39 USD
76992-238 76992-240 76992-242
tert-Butyl (2-amino-2-iminoethyl)carbamate hydrochloride 95%
tert-Butyl (2-amino-2-iminoethyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClN₃O₂
MDL Number: MFCD12913031
CAS Number: 691898-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR