2,6-Difluoronicotinic acid 96%

Supplier: Ambeed
Warning

A627060-100G A627060-1G A627060-25G A627060-5G
77005-562EA 231 USD
77005-562 77005-564 77005-566 77005-568
2,6-Difluoronicotinic acid 96%

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₂
MDL Number: MFCD08064047
CAS Number: 171178-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR