2-Methylindol-3-ylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylindole-3-acetic acid

A626523-10G A626523-1G A626523-25G A626523-5G
77005-548EA 48.81 USD
77005-548 77005-550 77005-552 77005-554
2-Methylindol-3-ylacetic acid 98%
2-Methylindol-3-ylacetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD00075006
CAS Number: 1912-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR