3-Pyrrol-1-ylpropionic acid 98%

Supplier: Ambeed
Warning

A625488-1G A625488-250MG A625488-25G A625488-5G
77035-308EA 23 USD
77035-308 77035-310 77035-312 77035-314
3-Pyrrol-1-ylpropionic acid 98%
3-Pyrrol-1-ylpropionic acid

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MDL Number: MFCD03844729
CAS Number: 89059-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR