Ethyl-1-boc-4-ethyl-4-piperidine carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-Boc-4-ethyl-4-piperidine carboxylate

A625313-1G A625313-250MG A625313-5G
76992-232EA 29 USD
76992-232 76992-234 76992-236
Ethyl-1-boc-4-ethyl-4-piperidine carboxylate 97%
Ethyl-1-boc-4-ethyl-4-piperidine carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₇NO₄
MDL Number: MFCD10565657
CAS Number: 188792-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR