Methyl 4-bromo-6-methylnicotinate 95%

Supplier: Ambeed
Warning

A625112-5G A625112-1G A625112-250MG
76992-230EA 234.46 USD
76992-230 77684-505 77684-506
Methyl 4-bromo-6-methylnicotinate 95%
Methyl 4-bromo-6-methylnicotinate

Precaución: For research use only.

Formula: C₈H₈BrNO₂
MW: 230.05 g/mol
MDL Number: MFCD07374918
CAS Number: 886372-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR