Fmoc-Cys(Mtt)-OH 97%

Supplier: Ambeed
Warning

A624873-1G A624873-25G A624873-5G
76992-226EA 25 USD
76992-226 76992-228 77891-235
Fmoc-Cys(Mtt)-OH 97%
Fmoc-Cys(Mtt)-OH

Precaución: for Research Use Only

Formula: C₃₈H₃₃NO₄S
MW: 599.745 g/mol
MDL Number: MFCD02259492
CAS Number: 269067-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR