7-Chloro-6-nitro-4-hydroxyquinazoline 98%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-6-nitroquinazolin-4(3H)-one, 7-Chloro-6-nitro-4(3H)-quinazolinone, 7-Chloro-6-nitro-4-quinazolinol, 7-Chloro-6-nitro-4-quinazolinone

A624527-100G A624527-1G A624527-25G A624527-5G
77035-300EA 214.31 USD
77035-300 77035-302 77035-304 77035-306
7-Chloro-6-nitro-4-hydroxyquinazoline 98%
7-Chloro-6-nitro-4-hydroxyquinazoline

Precaución: for Research Use Only

Formula: C₈H₄ClN₃O₃
MW: 225.59 g/mol
MDL Number: MFCD09388771
CAS Number: 53449-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR