4-(1,3,2-dioxaborinan-2-yl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxyphenylboronic acid propanediol ester

A623862-1G A623862-250MG A623862-5G
77035-288EA 16 USD
77035-288 77035-290 77035-292
4-(1,3,2-dioxaborinan-2-yl)benzoic acid 97%
4-(1,3,2-dioxaborinan-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BO₄
MDL Number: MFCD06203595
CAS Number: 126747-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR