5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%

Supplier: Ambeed
Danger

Synonyms: 5-Chloro-2-methoxypyridine-3-boronic acid pinacol ester

A623170-100MG A623170-1G A623170-250MG
77035-274EA 37.14 USD
77035-274 77035-276 77035-278
5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%
5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₃
MW: 269.54 g/mol
MDL Number: MFCD09878898
CAS Number: 1083168-96-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR