6-Chloro-5-fluoropyridine-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A622910-1G A622910-250MG
77035-270EA 78 USD
77035-270 77035-272
6-Chloro-5-fluoropyridine-3-carbaldehyde 98%
6-Chloro-5-fluoropyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₃ClFNO
MDL Number: MFCD13189060
CAS Number: 950691-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR