Methyl-3-nitrophenylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Nitrophenylacetate

A622431-100MG A622431-1G A622431-250MG A622431-25G A622431-5G
77035-256EA 15.92 USD
77035-256 77035-258 77035-260 77035-262 77035-264
Methyl-3-nitrophenylacetate 98%
Methyl-3-nitrophenylacetate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.18 g/mol
MDL Number: MFCD08669939
CAS Number: 10268-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR