5-Bromo-2-chloro-4-iodopyridine 98%

Supplier: Ambeed
Warning

A622072-1G A622072-25G A622072-5G
77005-518EA 10 USD
77005-518 77005-520 77005-522
5-Bromo-2-chloro-4-iodopyridine 98%
5-Bromo-2-chloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClIN
MDL Number: MFCD09972189
CAS Number: 401892-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR