4-Methyl-7-nitro-1H-indazole 97+%

Supplier: Ambeed
Danger

A621277-100MG A621277-1G A621277-250MG
77005-512EA 24.41 USD
77005-512 77005-514 77005-516
4-Methyl-7-nitro-1H-indazole 97+%
4-Methyl-7-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.1601 g/mol
MDL Number: MFCD08689688
CAS Number: 104103-06-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR