Ethyl 4-chloro-2-methylbenzoate 98+%

Supplier: Ambeed
Warning

A620985-100G A620985-25G A620985-5G
77018-126EA 182.48 USD
77018-126 77018-128 77018-130
Ethyl 4-chloro-2-methylbenzoate 98+%
Ethyl 4-chloro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.64 g/mol
MDL Number: MFCD11554963
CAS Number: 15393-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR