3-Fluoro-4-piperidinone hydrochloride 98%

Supplier: Ambeed
Warning

A620901-5G A620901-1G A620901-250MG
77018-124EA 115 USD
77018-124 77891-232 77891-233
3-Fluoro-4-piperidinone hydrochloride 98%
3-Fluoro-4-piperidinone hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClFNO
MDL Number: MFCD11506659
CAS Number: 1070896-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR