2-Amino-4-chloro-N-methylbenzamide 95%

Supplier: Ambeed
Warning

A620184-1G A620184-250MG A620184-5G
77018-116EA 69 USD
77018-116 77018-118 77018-120
2-Amino-4-chloro-N-methylbenzamide 95%
2-Amino-4-chloro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O
MDL Number: MFCD09045571
CAS Number: 104775-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR