Ethyl-3-phenylpropionate 98%

Supplier: Ambeed
Warning

A619197-1000G A619197-100G A619197-25G A619197-500G
77005-504EA 106 USD
77005-504 77005-506 77005-508 77005-510
Ethyl-3-phenylpropionate 98%
Ethyl-3-phenylpropionate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MDL Number: MFCD00009206
CAS Number: 2021-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR