(3-Methylquinolin-2-yl)methanamine dihydrochloride 95+%

Supplier: Ambeed
Warning

A618592-100MG A618592-1G A618592-250MG
77018-098EA 56 USD
77018-098 77018-100 77018-102
(3-Methylquinolin-2-yl)methanamine dihydrochloride 95+%
(3-Methylquinolin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄Cl₂N₂
MDL Number: MFCD27923291
CAS Number: 1956322-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR