3-Chloro-6-isopropoxypyridazine 98%

Supplier: Ambeed
Danger

A618306-5G A618306-1G A618306-100MG A618306-250MG
77005-502EA 97 USD
77005-502 77673-668 77882-445 77882-446
3-Chloro-6-isopropoxypyridazine 98%
3-Chloro-6-isopropoxypyridazine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
MDL Number: MFCD00270293
CAS Number: 3184-71-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR