O-Acetyl-N-tert-butylhydroxylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: O-Acetyl-N-(tert-butyl)hydroxylamine hydrochloride

A618284-1G A618284-250MG A618284-5G
76992-190EA 83.82 USD
76992-190 76992-192 76992-194
O-Acetyl-N-tert-butylhydroxylamine hydrochloride 97%
O-Acetyl-N-tert-butylhydroxylamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 167.63 g/mol
MDL Number: MFCD12545957
CAS Number: 851074-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR