(R)-1-(3-Chloro-4-methylphenyl)ethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A617253-100MG A617253-250MG
76992-182EA 27 USD
76992-182 76992-184
(R)-1-(3-Chloro-4-methylphenyl)ethanamine hydrochloride 97%
(R)-1-(3-Chloro-4-methylphenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃Cl₂N
MDL Number: MFCD16293365
CAS Number: 856562-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR