Methyl 2-chlorobenzo[d]thiazole-6-carboxylate 98+%

Supplier: Ambeed
Warning

A616829-100MG A616829-1G A616829-250MG A616829-5G
77035-230EA 43.5 USD
77035-230 77035-232 77035-234 77035-236
Methyl 2-chlorobenzo[d]thiazole-6-carboxylate 98+%
Methyl 2-chlorobenzo[d]thiazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂S
MW: 227.66 g/mol
MDL Number: MFCD13190216
CAS Number: 90792-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR