(S)-2,6-Diaminohexanoic acid (S)-2-aminosuccinic acid salt 98+%

Supplier: Ambeed
Warning

Synonyms: H-Lys-OH H-Asp-OH salt

A616505-100G A616505-25G A616505-5G
77005-486EA 8.49 USD
77005-486 77005-488 77005-492
(S)-2,6-Diaminohexanoic acid (S)-2-aminosuccinic acid salt 98+%
(S)-2,6-Diaminohexanoic acid (S)-2-aminosuccinic acid salt

Precaución: for Research Use Only

Formula: C₁₀H₂₁N₃O₆
MW: 279.29 g/mol
MDL Number: MFCD00058085
CAS Number: 27348-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR